Bis(2-ethylhexyl)phosphate ≥95%

Leverandør: Thermo Fisher Scientific
Danger

17723.22 17723.36 17723.A3
17723.22EA 570 NOK
17723.22 17723.36 17723.A3
Bis(2-ethylhexyl)phosphate ≥95%
Bis(2-ethylhexyl)phosphate

Purity: 95% CAS No.: 298-07-7 Molecular Formula: C16H35O4P Formula Weight: 322.43 Flash Point: 171°(340°F) Physical Form: Liquid Density: 0.965 Refractive Index: 1.4420 Appearance: Colourless pH: 2-3 Melting Point: -50° Boiling Point: 48°/12mm UN No.: 1902 MDL No.: MFCD00009492


Extraction of metals

Formel: C₁₆H₃₅O₄P
Kokepunkt: 48 °C (12 mmHg)
Smeltepunkt: –50 °C
Tetthet: 0,965
Flash punkt: 171 °C (340 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00009492
CAS-nummer: 298-07-7
EINECS: 206-056-4
FN: 1902
ADR: 8,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR