Kobber(II)klorid dihydrat, BAKER ANALYZED® ACS, J.T.Baker®

Leverandør: Avantor
Warning

Synonymer: Copper dichloride dihydrate, Cupric chloride dihydrate

1792-01EA 5650 NOK
1792-01
Kobber(II)klorid dihydrat, BAKER ANALYZED® ACS, J.T.Baker®
Kobber(II)klorid dihydrat
Formel: CuCl₂·2H₂O
MW: 170.48 g/mol
Smeltepunkt: 100
Tetthet: 3.054 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149674
CAS-nummer: 10125-13-0
EINECS: 231-210-2
FN: 2802
ADR: 8 III
Merck Indeks: 13,02660

Bestill nå

Spesifikasjon Testresultater

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use 99.0 - 101.0 %
ACS - Assay (CuCl₂2H₂O) 99.0 - 101.0 %
ACS - Insoluble Matter ≤ 0.01 %
ACS - pH of 5% Solution at 25°C 2.0 - 4.0
ACS - Nitrate (NO₃) ≤ 0.015 %
ACS - Sulfate (SO₄) ≤ 0.005 %
ACS - Calcium (Ca) ≤ 0.005 %
ACS - Iron (Fe) ≤ 0.005 %
ACS - Nickel (Ni) ≤ 0.01 %
ACS - Potassium (K) ≤ 0.01 %
ACS - Sodium (Na) ≤ 0.02 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR