Sample loop kit
Leverandør: Cytiva
18-0404-01
18-0404-01EA 5550 NOK
18-0404-01
Sample loop kit
Kromatografiutstyr Kromatografiprøvesløyfer
Each sample loop consists of tubing 1,8 mm OD and two tubing connectors (M6).
Leveringsinformasjon: The kit contains: 1×25 µl sample loop 0,5 mm ID; 1×50 µl sample loop 0,5 mm ID; 1×100 µl sample loop 0,5 mm ID; 1×200 µl sample loop 0,8 mm ID; 1×500 µl sample loop 0,8 mm ID.