Naphthol AS-BI phosphate solution_SC 4 g/L methanol Stabilised, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: 7-Bromo-3-hydroxy-2-naphthoic-o-anisidide phosphate, Naphthol AS-BI Phosphoric acid solution, Naphthol AS-BI-Phosphate, 6-Bromo-3-((2-methoxyphenyl)carbamoyl)naphthalen-2-yl dihydrogen phosphate

MQ500

1802-50ML
1802-50MLEA 1170 NOK
1802-50ML
Naphthol AS-BI phosphate solution_SC 4 g/L methanol Stabilised, Sigma-Aldrich®
Naphthol AS-BI phosphate

Naphthol AS-BI Phosphoric acid solution for use in the Sigma Lymphocyte Enzyme (Acid Phosphatase) Procedure, No. 181A.


  • Suitable for haematology
  • Suitable for histology

Formel: C₁₈H₁₅BrNO₆P
MW: 452.1926 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00004058
CAS-nummer: 1919-91-1
FN: 1993
ADR: 3 III
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR