Phosphate Buffer Solution

Leverandør: SIGMA ALDRICH MICROSCOPY

1805-1VL
1805-1VLEA 931 NOK
1805-1VL
Phosphate Buffer Solution
Buffere Buffere, biologiske

Contains sodium and potassium phosphates. Intended for use in the Sigma Butyrate Esterase Procedure, No. 181B.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
pH 7,60 - 7,80 (25 °C)
Skjema Powder
Lagringstemp RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR