VWR® SD 820, Membranpumpe, selvtørkende, for fuktige gasser
Leverandør: VWR Collection
181-0094EA 28400 NOK
181-0094
VWR® SD 820, Membranpumpe, selvtørkende, for fuktige gasser
Pumper Membranpumper
Dual-head, dry running diaphragm pump with an integrated self-drying system. There is a wide range of applications for this pump in laboratories, especially whenever clean vacuum is required and moist gases must be pumped down. Examples include operation of vacuum drying chambers (for drying or heat-treating substances and components) or steam sterilisers for sterilising instruments, vessels, filters and textiles. This pump can be used in a wide range of applications due to the pump's high resistance to aggressive media. The pump includes a PTFE coated diaphragm with patented tensionMore Product Information
Bestill nå
SPESIFIKASJONER
Modell SD 820
Environmentally Preferable
Gjennomstrømningshastighet 20
Overtrykk ved drift (bar) 1
Sluttrykk (abs.) (mbar) 10 mbar
Beskyttelsesklasse IP 44
B×D×H 312×220×177 mm
Vekt 9,6 kg
Sluttvakuum (abs.) 10 mbar