VWR®, Peristaltic Pump Head
Leverandør: VWR Collection
181-0291EA 3875 NOK
181-0291
VWR®, Peristaltic Pump Head
Pumper Pumpehoder Peristaltiske pumpehoder
The pump heads are rugged fabrications of polyamide, acetal and stainless steel with an integral pump head drive shaft with dual ball race bearings.
Maximum head: 20 m
Leveringsinformasjon: The pumps are supplied with a stainless steel mounting plate.