Cleanroom vacuum cleaner, EMI 1000
Leverandør: TIGER-VAC
110133B
181-0341EA 42375 NOK
181-0341
Cleanroom vacuum cleaner, EMI 1000
Vakuumrensere
Designed for general clean up in contamination controlled environments, cleanroom compatible and ideal for pharmaceutical industry. Designed for safe collection, containment and disposal of potent compounds. Construction: Stainless steel AISI 316 2B.
  • Cleanroom compatible with ISO Class 4 (former Class 10)
  • Static-free and ESD safe
  • five-stage filtration system include an ULPA filter U15, HEPA filter H14, cloth filter, safety filter and disposable filter bag
  • Single phase through-flow motor
  • Dual speed control to regulate suction
  • Filter chamber and recovery