e.jet pump, Microsart®
Leverandør: Sartorius
166MP-4
181-0589EA 34825 NOK
181-0589
e.jet pump, Microsart®
Pumper Vakuumpumper
The Microsart® e.jet is a laboratory vacuum pump which creates vacuum and concomitantly transfers the filtered liquid to waste.
  • For vacuum filtration in microbiological analysis
Microsart® e.jet is ideal for sample preparation in microbiological analysis achieving a trans membrane pressure of 600 mbar and a flow rate of 4,0 Nl/min (4,0 Normlitre water displacement by air in 1 minute). Constant flow rates and a defined maximum vacuum guarantees smooth and reliable filtration.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Gjennomstrømningshastighet >4,0 Nl/min
Inngangs-/utgangskobling forbindelse Quick connection on hose nipples for DN 10 tubing
Maks. trykk 1,0 bar
Maks. vacuum 0,4 bar
Driftstemperatur +5 /+40 ˚C max. ambient temp.;+5 /+80 ˚C max. liquid temp.
Beskyttelsesklasse III (protection type IP 64)
B×D×H 120×170×190 mm
Vekt 1,425 g