Poly(Bisphenol A carbonate) analytical standards kit, molecular weight series, for GPC
Leverandør: Merck
181676-1SET
181676-1SETEA 5200 NOK
181676-1SET
Poly(Bisphenol A carbonate) analytical standards kit, molecular weight series, for GPC
Standarder kromatografi standarder
Poly(Bisphenol A carbonate) (PC) is a thermoplastic material. It is injected in molding operations and is majorly studied to understand its thermal behaviour, especially thermal degradation property.
Kit components:
Average Mw 23000; average Mn 13,000 25 g
Average Mw 31000; average Mn 18,000 25 g
Average Mw 40000; average Mn 21,000 25 g
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d78d4d8d8658650f71c5d5c2ec3e83d28dab796a&vid=d78d4d8d8658650f71c5d5c2ec3e83d28dab796a&prid=web&clid=SAPDAM