Polystyrene, average Mw 290000 (typical), average Mn 130000 (typical), analytical standard, Supelco®
Leverandør: Merck
182435-25G
182435-25GEA 1750 NOK
182435-25G
Polystyrene, average Mw 290000 (typical), average Mn 130000 (typical), analytical standard, Supelco®
Standarder kromatografi standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e831e62235cace0b7c8066d363c86accc38e475d&vid=e831e62235cace0b7c8066d363c86accc38e475d&prid=web&clid=SAPDAM