Poly(vinyl chloride), average Mw 85000 (Typical), average Mn 40000 (Typical), analytical standard, Supelco®
Leverandør: Merck
182621-25G
182621-25GEA 1550 NOK
182621-25G
Poly(vinyl chloride), average Mw 85000 (Typical), average Mn 40000 (Typical), analytical standard, Supelco®
Standarder kromatografi standarder
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eac198c82137da8fd3725a74cd6e189a68e40f0a&vid=eac198c82137da8fd3725a74cd6e189a68e40f0a&prid=web&clid=SAPDAM
Bestill nå
SPESIFIKASJONER
CAS-nr 9002-86-2