Supelco® TraceCERT® Organic Reference Standard, Metrafenone
Leverandør: Merck
18996-50MG
18996-50MGEA 1275 NOK
18996-50MG
Supelco® TraceCERT® Organic Reference Standard, Metrafenone
Standarder Organiske standarder
Metrafenone CRM may also be used in the residue analysis of: 209 pesticides and persistent organic pollutants (POPs) extracted by QuEChERS-based procedure from fresh bovine and chicken liver samples using gas chromatography coupled to tandem mass spectrometry (GC-MS/MS). 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry. 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS). 379 pesticides in human serum samples extracted by QuEChERS method using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Metrafenone in nine fresh vegetable samples using matrix solid-phase dispersion (MSPD) and HPLC-UV. Metrafenone in bitter gourd and soil samples using gas chromatography combined with an electron capture detector (GC-ECD).
Sertifiseringer: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
NB: P273 - P391 - P501