Kalsiumklorid 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution

Leverandør: VWR Chemicals
Warning

Synonymer: Chloro calcium, Calcium dichloride, CaCl2

190464KEA 2050 NOK
190464K
Kalsiumklorid 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution
Kalsiumklorid
Formel: CaCl₂
MW: 110.98 g/mol
Kokepunkt: 1600
Smeltepunkt: 772
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010903
CAS-nummer: 10043-52-4
EINECS: 233-140-8
Merck Indeks: 14,01659

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.998 - 1.002 mol/l
NIST traceable Confirmed

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR