Natriumtetrahydroborat 0.5 M in diglyme, AcroSeal®

Leverandør: Thermo Fisher Scientific
Danger

191131000
191131000.EA 1025 NOK
191131000.
Natriumtetrahydroborat 0.5 M in diglyme, AcroSeal®
Sodium borohydride
Formel: NaBH₄
Tetthet: 0,945 g/cm³
Flash punkt: 69 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003518
CAS-nummer: 16940-66-2
ADR: 4.3,I

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Solution
Strength 0.45 to 0.55 M
Solution contains 18.915 g NaBH4/l C6H14O3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR