Sølvnitrat 0.02 mol/l (0.02 N) aqueous solution, AVS TITRINORM Volumetric

Leverandør: VWR Chemicals

Synonymer: Lunar caustic, Silver mononitrate, Nitric acid silver salt

191265JEA 1005 NOK
191265J
Sølvnitrat 0.02 mol/l (0.02 N) aqueous solution, AVS TITRINORM Volumetric
Sølvnitrat
Formel: AgNO₃
MW: 169.87 g/mol
Kokepunkt: 444
Smeltepunkt: 210
Tetthet: 1.01 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003414
CAS-nummer: 7761-88-8
EINECS: 231-853-9
FN: 3082
ADR: 9 III
Merck Indeks: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Titer (20°C; real value 0.2 % accuracy) 0.0198 - 0.0202 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR