Vanadomolybdate reagent

Leverandør: VWR Chemicals
Danger

191844WEA 1575 NOK
191844W
Vanadomolybdate reagent
Vanadomolybdate reagent
Kokepunkt: ~100 °C (1013 hPa)
Tetthet: 1.2 g/cm³ (20 °C)
FN: 2031
ADR: 8 II

Bestill nå

Spesifikasjon Testresultater

Colour reaction Satisfactory


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR