Mordant Blue 9 50% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: 6-(5-Chloro-2-hydroxy-4-sulphonphenylazo)-5-hydroxy-1-naphthalene-sulphonic acid sodium salt, 6-(5-Chloro-2-hydroxy-3-sulphophenylazo)-5-hydroxy-1-naphthalenesulphonic acid disodium salt

195219-25G
195219-25GEA 512 NOK
195219-25G
Mordant Blue 9 50% (dye content), Sigma-Aldrich®
Mordant Blue 9

Mordant Blue 9, Dye content 50 % (6-(5-Chloro-2-hydroxy-3-sulfophenylazo)-5-hydroxy-1-naphthalenesulfonic acid disodium salt; 6-(5-Chloro-2-hydroxy-4-sulfonphenylazo)-5-hydroxy-1-naphthalene-sulfonic acid sodium salt)

Formel: C₁₆H₉ClN₂Na₂O₈S₂
MW: 502.81 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 3624-68-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81128b29082f38b2f7927909e936dbcbb988f3b1&vid=1780fcf306b17352c48d2d8402d84f0d3530411d&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR