Natriumazid ≥99.7%, OR, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials
Danger

1953-09EA 16025 NOK
1953-09 1953-59
Natriumazid ≥99.7%, OR, Macron Fine Chemicals™
Sodium azide

Macron Fine Chemicals™ OR: organic reagents that are suitable for research applications.

Formel: NaN₃
MDL nummer: MFCD00003536
CAS-nummer: 26628-22-8
FN: 1687
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

For Laboratory,Research,or Manufacturing Use = 99.7 %
Assay (NaN₃) ≥99.7 %
Loss on Drying at 105°C ≤0.1 %
Typical property (not a specification): Appearance (white, crystalline
powder)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR