Supelco® Organic Reference Standard, Geranyl Pyrophosphate Lithium Salt

Leverandør: Merck

19533-5MG
19533-5MGEA 3925 NOK
19533-5MG
Supelco® Organic Reference Standard, Geranyl Pyrophosphate Lithium Salt
Standarder Organiske standarder

Forpakning: Bottomless glass bottle. Contents are inside inserted fused cone.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e34b1fce0a5ef303367cda65269303d159facb4&vid=0e34b1fce0a5ef303367cda65269303d159facb4&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Synonym(s) (E)-3,7-Dimethyl-2,6-octadien-1-ol trihydrogen pyrophosphate lithium salt; (E)-3,7-Dimethyl-2,6-octadien-1-yl pyrophosphate lithium salt; GPP-Li
Environmentally Preferable
M-Clarity quality level MQ100
CAS-nr 21141-43-5
Formel C₁₀H₂₀O₇P₂ ·xLi+
Molekylvekt 314,21 (free acid basis)
Flammepunkt Not applicable
Fraktforhold DICE
Lagringsforhold –20 °C
Lagringstemp −20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR