Erythrosin B disodium salt, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: C.I. 45430

M-Clarity™ quality level = MQ200

198269-25G
198269-25GEA 525 NOK
198269-25G
Erythrosin B disodium salt, Sigma-Aldrich®
Erythrosin B disodium salt
Formel: C₂₀H₆I₄Na₂O₅
MW: 879.86 g/mol
MDL nummer: MFCD00144257
CAS-nummer: 16423-68-0
EINECS: 240-474-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84be5218cd5911af8593cca43308b4f3414962ff&vid=1949387928e82fe712a333fa1ac27fe2b775478e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR