Resazurin sodium salt, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
Warning
M-Clarity™ quality level = MQ200
199303-1G 199303-25G 199303-5G
199303-1GEA 356 NOK
199303-1G 199303-25G 199303-5G
Resazurin sodium salt, Sigma-Aldrich®
Resazurin sodium salt
Formel: C₁₂H₆NNaO₄
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005036
CAS-nummer: 62758-13-8
EINECS: 263-718-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa7d92ac93005222e575530afa8e86bd5a79064e&vid=fc1000dd6dc4e3b80d5d52e3a07862ccf7290ed3&prid=web&clid=SAPDAM