Sudan black B, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

199664-25G
199664-25GEA 769 NOK
199664-25G
Sudan black B, Sigma-Aldrich®
Sudan black B
Formel: C₂₉H₂₄N₆
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006919
CAS-nummer: 4197-25-5
EINECS: 224-087-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=077ea042ad15e641f4f3bcb718d6b45a28db36cc&vid=077ea042ad15e641f4f3bcb718d6b45a28db36cc&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR