Acid Red 18 (Food Red No. 102), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: Cochineal Red A, New Coccine, Scarlet 3R, Sodium 7-hydroxy-8-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1,3-disulphonate, Ponceau 4R, Cochenille red

199737-25G
199737-25GEA 439 NOK
199737-25G
Acid Red 18 (Food Red No. 102), Sigma-Aldrich®
Acid Red 18 (Food Red No. 102)

New Coccine has been used to determine its decolorization by fungi Phanerochaete chrysosporium and Pleurotus sajorcaju.

Formel: C₂₀H₁₄N₂O₁₀S₃
MW: 604.46 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004084
CAS-nummer: 2611-82-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24abb47bd790ea036e151eae66224dfc591911a7&vid=bf9fbf08a7128b7b96329723cd5db9d19c8a956e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR