Atorvastatin Ethyl Ester, Pharmaceutical Analytical Impurities

Leverandør: USP

1A00010
1A00010.EA 8350 NOK
1A00010.
Atorvastatin Ethyl Ester, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse Atorvastatin Ethyl Ester
Synonym(s) ethyl (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate
Environmentally Preferable
CAS-nr 1146977-93-6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR