Desmethoxy Omeprazole, Pharmaceutical Analytical Impurities

Leverandør: USP

1A00060
1A00060.EA 9275 NOK
1A00060.
Desmethoxy Omeprazole, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse Desmethoxy Omeprazole
Synonym(s) 2-(((3,5-dimethylpyridin-2-yl)methyl)sulFood Ingredientsnyl)-5-methoxy-1H-benzo[d]imidazole
Environmentally Preferable
CAS-nr 110374-16-8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR