Ibuprofen Tetraethylene Glycol Diester, Pharmaceutical Analytical Impurities

Leverandør: USP

1A00480
1A00480.EA 5600 NOK
1A00480.
Ibuprofen Tetraethylene Glycol Diester, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse Ibuprofen Tetraethylene Glycol Diester
Synonym(s) ((oxybis(ethane-2,1-diyl))bis(oxy))bis(ethane-2,1-diyl) bis(2-(4-isobutylphenyl)propanoate))
Environmentally Preferable
CAS-nr 1161014-75-0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR