Risperidone Difluoroketone, Pharmaceutical Analytical Impurities
Leverandør: USP
1A00600
1A00600.EA 6775 NOK
1A00600.
Risperidone Difluoroketone, Pharmaceutical Analytical Impurities
Standarder Organiske standarder
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.
Bestill nå
SPESIFIKASJONER
Beskrivelse Risperidone Difluoroketone
Synonym(s) 3-(2-(4-(2,4-difluorobenzoyl)piperidin-1-yl)ethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
Environmentally Preferable
CAS-nr 158697-67-7