Pemetrexed Dimethyl Ester, Pharmaceutical Analytical Impurities

Leverandør: USP

1A00640
1A00640.EA 5175 NOK
1A00640.
Pemetrexed Dimethyl Ester, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse Pemetrexed Dimethyl Ester
Synonym(s) dimethyl (4-(2-(2-amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzoyl)-L-glutamate
Environmentally Preferable
CAS-nr 155405-81-5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR