Alpha-Naphthol, Pharmaceutical Analytical Impurities

Leverandør: USP

1A01070
1A01070.EA 12650 NOK
1A01070.
Alpha-Naphthol, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse Alpha-Naphthol
Synonym(s) Naphthalen-1-ol
Environmentally Preferable
CAS-nr 90-15-3
UN No. 2811

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR