Tolterodine Dimer, Pharmaceutical Analytical Impurities
Leverandør: USP
1A01620
1A01620.EA 19075 NOK
1A01620.
Tolterodine Dimer, Pharmaceutical Analytical Impurities
Standarder Organiske standarder
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.
Bestill nå
SPESIFIKASJONER
Beskrivelse Tolterodine Dimer
Environmentally Preferable
Synonym(s) 2,2'-((isopropylazanediyl)bis(1-phenylpropane-3,1-diyl))bis(4-methylphenol)
CAS-nr 854306-72-2