3-Hydroxy Desloratadine, Pharmaceutical Analytical Impurities
Leverandør: USP
1A01650
1A01650.EA 19075 NOK
1A01650.
3-Hydroxy Desloratadine, Pharmaceutical Analytical Impurities
Standarder Organiske standarder
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.
Bestill nå
SPESIFIKASJONER
Beskrivelse 3-Hydroxy Desloratadine
Synonym(s) 8-chloro-11-(piperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridin-3-ol
Environmentally Preferable
CAS-nr 119410-08-1