N-Benzylacetamide, Pharmaceutical Analytical Impurities

Leverandør: USP

1A01810
1A01810.EA 11175 NOK
1A01810.
N-Benzylacetamide, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse N-Benzylacetamide
Synonym(s) N-Acetylbenzylamine
Environmentally Preferable
CAS-nr 588-46-5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR