Isobutylglutarmonoamide, Pharmaceutical Analytical Impurities

Leverandør: USP

1A01840
1A01840.EA 9525 NOK
1A01840.
Isobutylglutarmonoamide, Pharmaceutical Analytical Impurities
Standarder Organiske standarder

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Bestill nå

SPESIFIKASJONER

Beskrivelse Isobutylglutarmonoamide
Synonym(s) 3-(2-Amino-2-oxoethyl)-5-methylhexanoic Acid
Environmentally Preferable
CAS-nr 181289-15-6

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR