VWR® RiboReady™ Color Micro RNA Ladder
Leverandør: VWR Chemicals
1B1491-25RXNEA 17875 NOK
1B1491-25RXN
VWR® RiboReady™ Color Micro RNA Ladder
Elektroforese reagenser Molekylvekt markører
RiboReady™ Color Micro RNA ladder allows precise sizing of RNA molecules in denaturing acrylamide gels. The apparent sizes of bands in the RiboReady™ Color Micro RNA ladder are 100, 75, 50, 40, 30 and 20 bases in length.
  • 6 bands ranging from 20 to 100 bases
  • Contains pre-stained nucleic acids that are visible during electrophoresis
Ready to use mixture that does not require heating or addition of a denaturing agent before use.