VWR® RiboReady™, Color RNA Ladder, 1 kb
Leverandør: VWR Chemicals
1B1492-25RXNEA 8575 NOK
1B1492-25RXN
VWR® RiboReady™, Color RNA Ladder, 1 kb
Elektroforese reagenser Molekylvekt markører
RiboReady™ Color 1 kb RNA ladder allows precise sizing of RNA molecules in denaturing acrylamide gels.
  • 6 bands ranging from 200 to 8000 bases
  • Ready to use mixture that does not require heating or addition of a denaturing agent before use
RiboReady™ Color 1 kb RNA ladder is made for precise sizing of RNA molecules in denaturing agarose gels. RiboReady™ Color 1 kb RNA ladder has the added advantage of containing pre-stained single-strand nucleic acids that are visible during electrophoresis. The apparent sizes of bands in the RiboReady™ Color 1 kb RNA ladder are 200, 500, 1000, 2000, 4000 and 8000 bases in length.