VWR® YPD, Media, Liquid, for Biotechnology
Leverandør: VWR Chemicals
1B1493-1LEA 3050 NOK
1B1493-1L
VWR® YPD, Media, Liquid, for Biotechnology
Mikrobiologiske medier Media, preparerte for mikrobiologi
A complete medium for the propagation and maintenance of yeast. The composition of YPD is (w/v) 1% yeast extract, 2% peptone, and 2% D-glucose.
pH (25 °C): 6,14 to 7,14