DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)

Leverandør: VWR Chemicals

Synonymer: Thymonucleic acid sodium salt, DNA sodium salt

1B1509-25GEA 2525 NOK
1B1509-25G
DNA sodium salt (Deoxyribonucleic acid sodium salt) (from fish sperm)
DNA sodium salt (Deoxyribonucleic acid sodium salt)

DNA, salt, from fish sperm is an effective blocking agent used to increase signal-to-noise ratio for detection during hybridisation.

MDL nummer: MFCD00130921
CAS-nummer: 9007-49-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR