Azure A eosinate, powder, crystals or chunks, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

200212-25G
200212-25GEA 1250 NOK
200212-25G
Azure A eosinate, powder, crystals or chunks, Sigma-Aldrich®
Azure A eosinate
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012112
CAS-nummer: 62298-43-5
EINECS: 263-491-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9e8b7642d1b345e5a2c4e210893e26f312d2359&vid=a9e8b7642d1b345e5a2c4e210893e26f312d2359&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Wavelength 285 - 291 nm
Absorbance ≥0.3
Absorbance, c = 0.005 g/l; In Methanol ≥0.4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR