Azure B eosinate, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

200239-25G
200239-25GEA 1020 NOK
200239-25G
Azure B eosinate, Sigma-Aldrich®
Azure B eosinate
Formel: C₅H₃₈Br₄N₆O₅S₂
MW: 1186.62 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 62298-42-4
EINECS: 263-490-7

Bestill nå

Spesifikasjon Testresultater

Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
TLC Consistent with Past Lots
Solubility (Color) c = 1mg/ml, Acetic Acid Dark Blue
Wavelength c = 0.005 g/l in Methanol 635 - 641 nm
Extinction Coefficient ≥47500
Wavelength 241 - 247 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR