Capillary tube on foot
Leverandør: GLASWARENFABRIK KARL HECHT
41955010
201-0216EA 1100 NOK
201-0216
Capillary tube on foot
Lærings materiell Læremateriale for fysikk Mechanics Learning Activities
Five communicating glass tubes with different inner diameters. Used to show capillary forces and how they are affected by the inner diameter of the pipes.
Leveringsinformasjon: Comes with foot.