Drying tubes with standard ground cone, angled, Quickfit®
Leverandør: SciLabware
MF23/1 MF23/2 MF23/3 MF23/4
201-0217EA 700 NOK
201-0217 201-0218 201-0526 BIBBMF23/4
Drying tubes with standard ground cone, angled, Quickfit®
Rør Tørkerør
PYREX®, borosilicate glass with ground glass cone.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Designed for use with a desiccant such as calcium chloride.