Reaksjonskolbe, rund bunn, med flat flens, DURAN®
Leverandør: DWK Life Sciences
243956303 243957102 243957608 243958604 243959103 243998608 243999107 243977301
201-0377EA 3475 NOK
201-0377 201-0378 201-0379 201-0380 SCOT243959103 SCOT243998608 SCOT243999107 SCOT243977301
Reaksjonskolbe, rund bunn, med flat flens, DURAN®
Reaksjonskolber Reaksjonskolber, kuleformet
DURAN® borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat flange (DN 100) with groove for vacuum seal. Maximum operating pressure at 250 °C: 1,0 bar for 2000, 4000 and 6000 ml; 0,5 bar for 10000 ml.
Important: At a maximum operating temperature of 250 °C and at permissible operating pressure, the temperature difference in the glass wall of the flat flange reaction vessels may not exceed 30 °C.
NB: Viktig: VedMore Product Information