Pudderkolber
Leverandør: Lenz Laborglas GmbH & CO.KG
03043070 03043073 03043037 03043049 03043058
201-0628EA 2950 NOK
201-0628 LENZ03043073 LENZ03043037 201-0626 201-0627
Pudderkolber
Kolber Kolber, rotasjonsfordamper
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for rotary evaporators, for drying of powder-like substances. With center neck, conical joint and inscription label.