Tørkerør, bøyd
Leverandør: Lenz Laborglas GmbH & CO.KG
05337001 05337002 05337003 05337004
201-5311EA 565 NOK
201-5311 201-1166 201-1167 201-5312
Tørkerør, bøyd
Rør Tørkerør
Clear DURAN® borosilicate 3.3 glass. Angled at 75° with ground joint cone.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Suitable for use with a desiccant such as calcium chloride.