Glassrør
Leverandør: Lenz Laborglas GmbH & CO.KG
05081000
201-2610EA 100 NOK
201-2610
Glassrør
Rør Gassdistribusjonsrør
DURAN®, borosilikatglass 3.3, klar.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Hollow prevents boiling splashes.