Multiple adapters, Quickfit®
Leverandør: SciLabware
MA4/23
201-3002EA 2000 NOK
201-3002
Multiple adapters, Quickfit®
Destillasjonsutstyr Destillasjonsadaptere, flere halser
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With three necks. Two vertical and one angled at 45°.