Digestion tubes, plain neck
Leverandør: SciLabware
TT100/26 TT250/42
201-3050EA 2450 NOK
201-3050 201-3051
Digestion tubes, plain neck
Rør Digeston Rør
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Designed for use with block heaters.