Gas sample tubes
Leverandør: Lenz Laborglas GmbH & CO.KG
05680143 05680152 05680158 05680170
201-3424EA 2300 NOK
201-3424 201-3421 201-3422 201-3423
Gas sample tubes
Rør Gassprøvetakningsrør
Duran®, borosilicate glass, clear. Both ends with a 3 NS/14 one-way stop cock. Sampling tube with GL 14 thread.
  • White labelling area
  • Excellent thermal and chemical resistance
  • Connection on one end with capillary tube (i. Ø: 2,7 mm ; o. Ø: 8,0 mm) and on the other end with tube connector (o. Ø: 8,0 mm)
Gas collection tubes for removing, storing and transporting gas samples.
Sertifiseringer: DIN 12473-1