Soxhlet extractors, Quickfit®
Leverandør: SciLabware
EX5/32 EX5/43 EX5/53 EX5/55/100 EX5/63 EX5/83 EX5/55
201-8373EA 2275 NOK
201-8373 201-8374 201-8375 201-8377 201-8378 201-8380 BIBBEX5/55
Soxhlet extractors, Quickfit®
Ekstraktorer Soxhlet- og Soxhlet-typeuttrekkere
PYREX®, borosilicate glass.
For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent.
Sertifiseringer: Conforms to BS 2071